missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-tert-Butyl-3,5-dimethylaniline, 97%

Brand:  Alfa Aesar™ H55164.MD

Code : NEW

Additional Details : CAS Number : 110993-40-3 Weight : 0.00025kg

Quantity 250mg

 View more versions of this product

Product Code. 15401328

  • £16.64 / 250mg


Estimated Shipment date
from Supplier 07-02-2018
Add to basket

Specifications

Chemical Identifiers

N-tert-Butyl-3,5-dimethylaniline
110993-40-3
177.29
MFCD07440135

Specifications

N-tert-Butyl-3,5-dimethylaniline
69° to 70°C (0.75mmHg)
250mg
1.5190
97%
MFCD07440135
0.901g/mL
Colorless to brown
110993-40-3
Liquid
177.29
Videos
SDS