missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-tert-Butyl-3,5-difluorobenzamide, 97%

Brand:  Alfa Aesar™ H61228.03

Code : NEW

Additional Details : CAS Number : 223444-32-4 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15407738

  • £41.55 / 1g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

N-tert-Butyl-3,5-difluorobenzamide
213.23
14713394
223444-32-4
MFCD11223983

Specifications

N-tert-Butyl-3,5-difluorobenzamide
1g
Solid
213.23
14713394
White
223444-32-4
97%
MFCD11223983
Videos
SDS