missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-tert-Butyl-2-nitrobenzenesulfonamide, 97%

Brand:  Alfa Aesar™ H60077.03

Code : NEW

Additional Details : CAS Number : 363587-67-1 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15406548

  • £358.49 / 1g


Estimated Shipment date
from Supplier 06-03-2018
Add to basket

Specifications

Chemical Identifiers

N-tert-Butyl-2-nitrobenzenesulfonamide
363587-67-1
258.29
MFCD00450577

Specifications

N-tert-Butyl-2-nitrobenzenesulfonamide
1g
Solid
258.29
Pale orange
363587-67-1
97%
MFCD00450577
Videos
SDS