missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyloxy)succinimide, 97%, Thermo Scientific Chemicals

Catalog Number p-4228475
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
13139-12-3
C9H13NO5
215.21
VTGFSVGZCYYHLO-UHFFFAOYSA-N
boc-osu, tert-butyl 2,5-dioxopyrrolidin-1-yl carbonate, tert-butyl n-succinimidyl carbonate, n-tert-butoxycarbonyloxy succinimide, tert-butyl succinimido carbonate, 1-tert-butoxycarbonyl oxy pyrrolidine-2,5-dione, 2,5-pyrrolidinedione, 1-1,1-dimethylethoxy carbonyl oxy, carbonic acid, 1,1-dimethylethyl 2,5-dioxo-1-pyrrolidinyl ester, boc-osu, tert-butyl n-succinimidyl carbonate, n-tert-butoxycarbonyloxy succinimide boc-osu
83168
tert-butyl (2,5-dioxopyrrolidin-1-yl) carbonate
CC(C)(C)OC(=O)ON1C(=O)CCC1=O
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
13139-12-3
C9H13NO5
215.21
VTGFSVGZCYYHLO-UHFFFAOYSA-N
boc-osu, tert-butyl 2,5-dioxopyrrolidin-1-yl carbonate, tert-butyl n-succinimidyl carbonate, n-tert-butoxycarbonyloxy succinimide, tert-butyl succinimido carbonate, 1-tert-butoxycarbonyl oxy pyrrolidine-2,5-dione, 2,5-pyrrolidinedione, 1-1,1-dimethylethoxy carbonyl oxy, carbonic acid, 1,1-dimethylethyl 2,5-dioxo-1-pyrrolidinyl ester, boc-osu, tert-butyl n-succinimidyl carbonate, n-tert-butoxycarbonyloxy succinimide boc-osu
83168
tert-butyl (2,5-dioxopyrrolidin-1-yl) carbonate
CC(C)(C)OC(=O)ON1C(=O)CCC1=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 88°C to 91°C
Color White
Elemental Analysis 48.22 to 52.24% C, 6.25 to 6.77% N
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass Bottle
Quantity 5 g
Formula Weight 215.21
Percent Purity 97%
Physical Form Powder
Chemical Name or Material N-(tert-Butoxycarbonyloxy)succinimide
Show More Show Less
compliance-icons
Product Identifier
  • N-(tert-Butoxycarbonyloxy)succinimide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Respiratory sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'