missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyl)-L-valine N'-methoxy-N'-methylamide, 97%, Thermo Scientific™

Catalog Number 11965671
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
87694-52-8
C12H24N2O4
260.33
MFCD00151897
RRBFCGUIFHFYQK-VIFPVBQESA-N
s-2-boc-amino-n-methoxy-n,3-dimethylbutanamide, n-boc-l-valine n'-methoxy-n'-methylamide, n-tert-butoxycarbonyl-l-valine n'-methoxy-n'-methylamide, tert-butyl s-1-methoxy methyl amino-3-methyl-1-oxobutan-2-yl carbamate, tert-butyl n-1s-1-methoxy methyl carbamoyl-2-methylpropyl carbamate, boc-val-n och3 ch3, n-boc-l-valinen'-methoxy-n'-methylamide, n-alpha-t-butyloxycarbonyl-n-methyl-n-methoxy-l-valine amide, 2s-2-tert-butoxy carbonylamino-n-methoxy-3-methyl-n-methylbutanamide
10083961
tert-butyl N-[(2S)-1-[methoxy(methyl)amino]-3-methyl-1-oxobutan-2-yl]carbamate
CON(C)C(=O)[C@@H](NC(=O)OC(C)(C)C)C(C)C
This item is not returnable. View return policy
87694-52-8
C12H24N2O4
260.33
MFCD00151897
RRBFCGUIFHFYQK-VIFPVBQESA-N
s-2-boc-amino-n-methoxy-n,3-dimethylbutanamide, n-boc-l-valine n'-methoxy-n'-methylamide, n-tert-butoxycarbonyl-l-valine n'-methoxy-n'-methylamide, tert-butyl s-1-methoxy methyl amino-3-methyl-1-oxobutan-2-yl carbamate, tert-butyl n-1s-1-methoxy methyl carbamoyl-2-methylpropyl carbamate, boc-val-n och3 ch3, n-boc-l-valinen'-methoxy-n'-methylamide, n-alpha-t-butyloxycarbonyl-n-methyl-n-methoxy-l-valine amide, 2s-2-tert-butoxy carbonylamino-n-methoxy-3-methyl-n-methylbutanamide
10083961
tert-butyl N-[(2S)-1-[methoxy(methyl)amino]-3-methyl-1-oxobutan-2-yl]carbamate
CON(C)C(=O)[C@@H](NC(=O)OC(C)(C)C)C(C)C
CAS Min % 96.0
CAS Max % 100.0
Melting Point 34°C to 37°C
Color White to Yellow
Boiling Point 248°C
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Specific Rotation -15
Formula Weight 260.34
Percent Purity 97%
Physical Form Liquid or Low Melting Solid
Chemical Name or Material N-(tert-Butoxycarbonyl)-L-valine N'-methoxy-N'-methylamide
Show More Show Less
Product Identifier
  • N-(tert-Butoxycarbonyl)-L-valine N'-methoxy-N'-methylamine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'