missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyl)-L-alanine N'-methoxy-N'-methylamide, 98%, ACROS Organics™

Brand:  Acros Organics 439430050

Code : 29

Additional Details : CAS Number : 87694-49-3 Weight : 0.05500kg


Quantity 5g

 View more versions of this product

Product Code. 11915671

  • £99.20 / 5g


Estimated Shipment
21-06-2018
Add to basket


Specifications

Chemical Identifiers

N-(tert-Butoxycarbonyl)-L-alanine N'-methoxy-N'-methylamide
97.5
≥97.5%
C10H20N2O4
87694-49-3
100.0
98%
232.28

Specifications

N-(tert-Butoxycarbonyl)-L-alanine N'-methoxy-N'-methylamide
97.5
Authentic
≥97.5%
C10H20N2O4
-25° to -29° (20°C, 589nm) (C=1 in methanol)
5g
87694-49-3
100.0
Crystalline Powder or Powder
98%
232.28
White to off-white
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • N-(tert-Butoxycarbonyl)-L-alanine N'-methoxy-N'-methylamide

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS