missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyl)-L-alanine N'-methoxy-N'-methylamide, 98%, Thermo Scientific™

Catalog Number 11995661
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
87694-49-3
C10H20N2O4
232.28
MFCD00151859
PWQIGBOSLQHOBT-UHFFFAOYNA-N
boc-ala-nme ome, s-tert-butyl 1-methoxy methyl amino-1-oxopropan-2-yl carbamate, boc-ala-n och3 ch3, tert-butyl n-1s-1-methoxy methyl carbamoyl ethyl carbamate, s-2-boc-amino-n-methoxy-n-methylpropanamide, n-tert-butoxycarbonyl-l-alanine n'-methoxy-n'-methylamide, n2-tert-butoxycarbonyl-n-methoxy-n-methyl-l-alaninamide, 2s-2-tert-butoxy carbonylamino-n-methoxy-n-methylpropanamide, s-tert-butyl 1-methoxy methyl amino-1-oxopropan-2-ylcarbamate, r-tert-butyl1-methoxy methyl amino-1-oxopropan-2-ylcarbamate
11806530
CON(C)C(=O)C(C)NC(=O)OC(C)(C)C
This item is not returnable. View return policy
87694-49-3
C10H20N2O4
232.28
MFCD00151859
PWQIGBOSLQHOBT-UHFFFAOYNA-N
boc-ala-nme ome, s-tert-butyl 1-methoxy methyl amino-1-oxopropan-2-yl carbamate, boc-ala-n och3 ch3, tert-butyl n-1s-1-methoxy methyl carbamoyl ethyl carbamate, s-2-boc-amino-n-methoxy-n-methylpropanamide, n-tert-butoxycarbonyl-l-alanine n'-methoxy-n'-methylamide, n2-tert-butoxycarbonyl-n-methoxy-n-methyl-l-alaninamide, 2s-2-tert-butoxy carbonylamino-n-methoxy-n-methylpropanamide, s-tert-butyl 1-methoxy methyl amino-1-oxopropan-2-ylcarbamate, r-tert-butyl1-methoxy methyl amino-1-oxopropan-2-ylcarbamate
11806530
CON(C)C(=O)C(C)NC(=O)OC(C)(C)C
CAS Min % 97.5
CAS Max % 100.0
Melting Point 143.5°C to 148.3°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Specific Rotation -25.5
IUPAC Name tert-butyl N-{1-[methoxy(methyl)carbamoyl]ethyl}carbamate
Formula Weight 232.28
Percent Purity 98%
Physical Form Crystalline Powder or Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)-L-alanine N'-methoxy-N'-methylamide
Show More Show Less
compliance-icons
Product Identifier
  • N-(tert-Butoxycarbonyl)-L-alanine N'-methoxy-N'-methylamide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'