missing translation for 'onlineSavingsMsg'
Learn More

N-[(S)-(+)-1-Ethoxycarbonyl-3-phenylpropyl]-L-alanine, 97%, Thermo Scientific™

Catalog Number p-126522
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
82717-96-2
C15H21NO4
279.34
MFCD00209976
CEIWXEQZZZHLDM-AAEUAGOBSA-N
s-2-s-1-ethoxy-1-oxo-4-phenylbutan-2-yl amino propanoic acid, n-s-1-ethoxycarbonyl-3-phenylpropyl-l-alanine, unii-10sz4ij77b, n-1 s-ethoxycarbonyl-3-phenylpropyl-l-alanine, n-s-+-1-ethoxycarbonyl-3-phenylpropyl-l-alanine, 2s-2-2s-1-ethoxy-1-oxo-4-phenylbutan-2-yl amino propanoic acid, s,s-n-ethoxy carbonyl-3-phenylpropyl alanine, n-s-1-carboxyethyl-l-homophenylalanine ethyl ester, s,s-n-1-ethoxycarbonyl-3-phenylpropyl alanine
5702571
CCOC(=O)[C@H](CCC1=CC=CC=C1)N[C@@H](C)C(O)=O
This item is not returnable. View return policy
82717-96-2
C15H21NO4
279.34
MFCD00209976
CEIWXEQZZZHLDM-AAEUAGOBSA-N
s-2-s-1-ethoxy-1-oxo-4-phenylbutan-2-yl amino propanoic acid, n-s-1-ethoxycarbonyl-3-phenylpropyl-l-alanine, unii-10sz4ij77b, n-1 s-ethoxycarbonyl-3-phenylpropyl-l-alanine, n-s-+-1-ethoxycarbonyl-3-phenylpropyl-l-alanine, 2s-2-2s-1-ethoxy-1-oxo-4-phenylbutan-2-yl amino propanoic acid, s,s-n-ethoxy carbonyl-3-phenylpropyl alanine, n-s-1-carboxyethyl-l-homophenylalanine ethyl ester, s,s-n-1-ethoxycarbonyl-3-phenylpropyl alanine
5702571
CCOC(=O)[C@H](CCC1=CC=CC=C1)N[C@@H](C)C(O)=O
CAS Min % 96.0
CAS Max % 100.0
Melting Point 148°C to 153°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 1 g
IUPAC Name (2S)-2-[[(2S)-1-ethoxy-1-oxo-4-phenylbutan-2-yl]amino]propanoic acid
Formula Weight 279.34
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material N-[(S)-(+)-1-Ethoxycarbonyl-3-phenylpropyl]-L-alanine, 97%
Show More Show Less
compliance-icons
Product Identifier
  • N-¢(S)-(+)-1-Ethoxycarbonyl-3-phenylpropyl!-L-alanine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'