missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Propyltrichlorosilane, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mL
1 L
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
CAS Min % | 97.5 |
CAS Max % | 100.0 |
Color | Colorless to Brown |
Density | 1.1900g/mL |
Boiling Point | 123.0°C to 124.0°C |
Flash Point | 31°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 97.5% min. (GC) |
Packaging | Glass bottle |
Linear Formula | CH3CH2CH2SiCl3 |
Show More |
Safety and Handling
Product Identifier
- n-Propyltrichlorosilane
- Danger
- Flammable liquid Category 3
- Skin corrosion/irritation Category 1 B
- H314-Causes severe skin burns and eye damage.
- H226-Flammable liquid and vapour.
- EUH014-Reacts violently with water.
- EUH029-Contact with water liberates toxic gas.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'