missing translation for 'onlineSavingsMsg'
Learn More

n-Propyltriacetoxysilane, 98%, Thermo Scientific™

Catalog Number 15427168
Click to view available options
Quantity:
250 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
CAS 17865-07-5
Quantity 250 mg
Molecular Formula C9H16O6Si
MDL Number MFCD00086768
Synonym propylsilanetriyl triacetate, propyltriacetoxysilane, n-propyltriacetoxysilane, silanetriol, propyl-, triacetate, silanetriol, 1-propyl-, 1,1,1-triacetate, bis acetyloxy propyl silyl acetate, triacetoxypropylsilane, propyl-silanetriotriacetate, propylsilanetriol triacetate, diacetyloxy propyl silyl acetate
InChI Key DKGZKEKMWBGTIB-UHFFFAOYSA-N
SMILES CCC[Si](OC(=O)C)(OC(=O)C)OC(=O)C
IUPAC Name [diacetyloxy(propyl)silyl] acetate
Molecular Weight (g/mol) 248.306
PubChem CID 87339
Formula Weight 248.31
Percent Purity 98%
Chemical Name or Material n-Propyltriacetoxysilane
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'