missing translation for 'onlineSavingsMsg'
Learn More

n-Propylboronic acid, 95%, ACROS Organics™

Brand:  Acros Organics 370870050

Code : 29

Additional Details : CAS Number : 17745-45-8 Weight : 0.05500kg


Quantity 5g
Packaging Glass bottle

 View more versions of this product

Product Code. 10531714

  • £111.70 / 5g


Estimated Shipment
15-06-2018
Add to basket


Specifications

Chemical Identifiers

n-Propylboronic acid
94.0
≥94%
C3H9BO2
87.91
17745-45-8
100.0
95%
CH3(CH2)2B(OH)2
MFCD01074564

Specifications

n-Propylboronic acid
17745-45-8
100.0
Glass bottle
≥94%
C3H9BO2
87.91
White to off-white
5g
94.0
Authentic
Crystalline Powder or Flakes
95%
CH3(CH2)2B(OH)2
MFCD01074564
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • n-Propylboronic acid

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS