missing translation for 'onlineSavingsMsg'
Learn More

Nω-Propyl-L-arginine hydrochloride, Tocris Bioscience™

Highly selective inhibitor of nNOS

Brand:  Tocris (Bio-Techne) 120050

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13428853

  • £717.36 / 50mg

Estimated Shipment date
from Supplier 30-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Highly selective and potent inhibitor of nNOS (Ki = 57 nM). Displays 3158-fold and 149-fold selectivity over iNOS and eNOS respectively. Hypotensive in vivo.
Specifications

Chemical Identifiers

2321366-46-3
216.285
n-omega-propyl-l-arginine, omega-propyl-l-arginine, n-?-propyl-l-arginine, 2s-2-amino-5-n'-propylcarbamimidoyl amino pentanoic acid, nomega-propyl-l-arginine, hddah inhibitor, 2b, nomegapropyl-l-arginine, tocris-1200, 3ar, d0o2ko
(2S)-2-amino-5-[(N'-propylcarbamimidoyl)amino]pentanoic acid
C9H20N4O2
AOMXURITGZJPKB-ZETCQYMHSA-N
447180
CCCN=C(N)NCCCC(C(=O)O)N

Specifications

Nω-Propyl-L-arginine hydrochloride
C9H20N4O2
AOMXURITGZJPKB-ZETCQYMHSA-N
(2S)-2-amino-5-[(N'-propylcarbamimidoyl)amino]pentanoic acid
447180
>98%
2321366-46-3
n-omega-propyl-l-arginine, omega-propyl-l-arginine, n-?-propyl-l-arginine, 2s-2-amino-5-n'-propylcarbamimidoyl amino pentanoic acid, nomega-propyl-l-arginine, hddah inhibitor, 2b, nomegapropyl-l-arginine, tocris-1200, 3ar, d0o2ko
CCCN=C(N)NCCCC(C(=O)O)N
216.285
252.74
50mg
Videos
Documents