missing translation for 'onlineSavingsMsg'
Learn More

n-Propyl isocyanate, 99%, ACROS Organics™

Brand:  Acros Organics 131605000

Code : 29

Additional Details : CAS Number : 110-78-1 Weight : 0.55000kg Transport : UN number : 2482 Chem class : 6.1 Pack group : I


Quantity 500g
Packaging Glass bottle

 View more versions of this product

Product Code. 10541381

  • £1314.60 / 500g


Call for latest availability
+44 (0)1509 555 500
Add to basket

Sea freight only: For EU stock allow up to 15 days delivery.

Specifications

Chemical Identifiers

n-Propyl isocyanate
110-78-1
100.0
99%
CH3CH2CH2NCO
MFCD00002045
99%
98.5
99%
C4H7NO
85.11

Specifications

n-Propyl isocyanate
99%
98.5
Authentic
1.3940 to 1.3970
Liquid
99%
Product may develop some turbidity or precipitate
85.11
0.8948g/mL
Clear yellow
−1°C
500g
110-78-1
100.0
Glass bottle
0.8948
99%
C4H7NO
CH3CH2CH2NCO
MFCD00002045
83.0° to 84.0°C
− 30.0°C
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • n-Propyl isocyanate

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Skin sensitiser Category 1

Hazard Statement
  • H318-Causes serious eye damage.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H314-Causes severe skin burns and eye damage.
  • H331-Toxic if inhaled.
  • H302-Harmful if swallowed.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

SDS