missing translation for 'onlineSavingsMsg'
Learn More

n-Propyl 3,4,5-trihydroxybenzoate, 98%, Thermo Scientific Chemicals

Catalog Number p-7081016
Click to view available options
Quantity:
5 g
100 g
500 g
2500 g
This item is not returnable. View return policy
121-79-9
C10H12O5
212.201
MFCD00002196
ZTHYODDOHIVTJV-UHFFFAOYSA-N
propyl gallate, n-propyl gallate, progallin p, tenox pg, nipagallin p, gallic acid, propyl ester, gallic acid propyl ester, nipa 49, benzoic acid, 3,4,5-trihydroxy-, propyl ester, 3,4,5-trihydroxybenzene-1-propylcarboxylate
4947
CHEBI:10607
propyl 3,4,5-trihydroxybenzoate
CCCOC(=O)C1=CC(=C(C(=C1)O)O)O
This item is not returnable. View return policy
121-79-9
C10H12O5
212.201
MFCD00002196
ZTHYODDOHIVTJV-UHFFFAOYSA-N
propyl gallate, n-propyl gallate, progallin p, tenox pg, nipagallin p, gallic acid, propyl ester, gallic acid propyl ester, nipa 49, benzoic acid, 3,4,5-trihydroxy-, propyl ester, 3,4,5-trihydroxybenzene-1-propylcarboxylate
4947
CHEBI:10607
propyl 3,4,5-trihydroxybenzoate
CCCOC(=O)C1=CC(=C(C(=C1)O)O)O

n-Propyl 3,4,5-trihydroxybenzoate is an anti-fade reagent in fluorescence microscopy to reduce photobleaching of fluorescent probes such as rhodamine and fluorescein. Also used as an antioxidant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
n-Propyl 3,4,5-trihydroxybenzoate is an anti-fade reagent in fluorescence microscopy to reduce photobleaching of fluorescent probes such as rhodamine and fluorescein. Also used as an antioxidant.

Solubility
Soluble in ethanol (50 mg/ml) and water (0.35 g/100 ml at 25°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY
Melting Point 146°C to 150°C
Quantity 5 g
Beilstein 1877976
Merck Index 147859
Solubility Information Soluble in ethanol (50mg/ml) and water (0.35g/100 ml at 25°C).
Formula Weight 212.2
Percent Purity 0.98
Chemical Name or Material n-Propyl 3,4,5-trihydroxybenzoate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'