missing translation for 'onlineSavingsMsg'
Learn More

N-(Propargyloxy)phthalimide, 98%, Thermo Scientific™

Catalog Number p-187597
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
4616-63-1
C11H7NO3
201.18
MFCD00005890
HBGZBVPXPDNXOV-UHFFFAOYSA-N
n-propargyloxy phthalimide, n-prop-2-ynyloxy phthalimide, 2-prop-2-yn-1-yloxy isoindoline-1,3-dione, n-propargyloxyphthalimide, 2-prop-2-yn-1-yloxy isoindole-1,3-dione, 2-prop-2-ynyloxybenzo c azoline-1,3-dione, acmc-20aka8, n-2-propynoxyphthalimide, n-2-propynyloxy phthalimide, phthalimide, n-2-propynyloxy
78357
2-prop-2-ynoxyisoindole-1,3-dione
C#CCON1C(=O)C2=CC=CC=C2C1=O
This item is not returnable. View return policy
4616-63-1
C11H7NO3
201.18
MFCD00005890
HBGZBVPXPDNXOV-UHFFFAOYSA-N
n-propargyloxy phthalimide, n-prop-2-ynyloxy phthalimide, 2-prop-2-yn-1-yloxy isoindoline-1,3-dione, n-propargyloxyphthalimide, 2-prop-2-yn-1-yloxy isoindole-1,3-dione, 2-prop-2-ynyloxybenzo c azoline-1,3-dione, acmc-20aka8, n-2-propynoxyphthalimide, n-2-propynyloxy phthalimide, phthalimide, n-2-propynyloxy
78357
2-prop-2-ynoxyisoindole-1,3-dione
C#CCON1C(=O)C2=CC=CC=C2C1=O
CAS Min % 97.5
CAS Max % 100.0
Melting Point 149°C to 152°C
Color Beige to Brown
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 1 g
Formula Weight 201.18
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material N-(Propargyloxy)phthalimide, 98%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'