missing translation for 'onlineSavingsMsg'
Learn More

N-Prop-2-ynylurea, Maybridge

Brand:  Maybridge CD11100EA

Code : 31

Additional Details : CAS Number : 5221-62-5 Weight : 0.06000kg

Quantity 10g
Packaging Amber Glass Bottle

 View more versions of this product

Product Code. 10743433

  • £326.00 / 10g


Estimated Shipment date
from Supplier 22-02-2018
Add to basket

Specifications

Chemical Identifiers

N-Prop-2-ynylurea
96.5
97%
C4H6N2O
98.1
5221-62-5
100.0
97%
H2NC(O)NHCH2Cidentical toCH
MFCD00052817

Specifications

N-Prop-2-ynylurea
5221-62-5
100.0
97%
C4H6N2O
98.1
10g
96.5
Amber Glass Bottle
97%
H2NC(O)NHCH2Cidentical toCH
MFCD00052817
Videos
SDS