missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Phthaloyl-L-phenylalanine, 98%

Brand:  Alfa Aesar™ H65025.06

Code : NEW

Additional Details : CAS Number : 5123-55-7 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15422449

  • £47.80 / 5g


Estimated Shipment date
from Supplier 23-05-2018
Add to basket

Description

Description

Applications
It is used as pharmaceutical intermediate.

Solubility
Soluble in methanol. (almost transparency)

Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

N-Phthaloyl-L-phenylalanine
295.29
90080
5123-55-7
MFCD00069738
N-Pht-Phe-OH; (S)-2-(1,3-Dioxo-2-isoindolinyl)-3-phenylpropionic acid

Specifications

N-Phthaloyl-L-phenylalanine
Powder
295.29
90080
White
5g
5123-55-7
98%
MFCD00069738
N-Pht-Phe-OH; (S)-2-(1,3-Dioxo-2-isoindolinyl)-3-phenylpropionic acid
181° to 185°C
Hygroscopic
Videos
SDS