missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Phthaloyl-L-phenylalanine, 98%, Thermo Scientific™
Click to view available options
Quantity:
5 g
25 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
It is used as pharmaceutical intermediate.
Solubility
Soluble in methanol. (almost transparency)
Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
It is used as pharmaceutical intermediate.
Solubility
Soluble in methanol. (almost transparency)
Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
Specifications
Melting Point | 181°C to 185°C |
Color | White |
Beilstein | 90080 |
Solubility Information | Soluble in methanol. (almost transparency) |
IUPAC Name | (2S)-2-(1,3-dioxoisoindol-2-yl)-3-phenylpropanoic acid |
Formula Weight | 295.29 |
Percent Purity | 98% |
Quantity | 25 g |
Sensitivity | Hygroscopic |
Physical Form | Powder |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'