missing translation for 'onlineSavingsMsg'
Learn More

N-Phenylrhodanine, 97+%, Thermo Scientific™

Catalog Number p-3593480
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1457-46-1
C9H7NOS2
209.29
MFCD00100196
DVRWEKGUWZINTQ-UHFFFAOYSA-N
3-phenylrhodanine, n-phenylrhodanine, 3-phenylrhodanin, rhodanine, 3-phenyl, 4-thiazolidinone, 3-phenyl-2-thioxo, 3-phenylrodanin, n-phenyl-rhodanine, 3-phenyl-2-thioxo-1,3-thiazolidin-4-one, 3-phenyl-2-thioxo-thiazolidin-4-one, 3-phenyl-rhodanin german
73822
3-phenyl-2-sulfanylidene-1,3-thiazolidin-4-one
C1C(=O)N(C(=S)S1)C2=CC=CC=C2
This item is not returnable. View return policy
1457-46-1
C9H7NOS2
209.29
MFCD00100196
DVRWEKGUWZINTQ-UHFFFAOYSA-N
3-phenylrhodanine, n-phenylrhodanine, 3-phenylrhodanin, rhodanine, 3-phenyl, 4-thiazolidinone, 3-phenyl-2-thioxo, 3-phenylrodanin, n-phenyl-rhodanine, 3-phenyl-2-thioxo-1,3-thiazolidin-4-one, 3-phenyl-2-thioxo-thiazolidin-4-one, 3-phenyl-rhodanin german
73822
3-phenyl-2-sulfanylidene-1,3-thiazolidin-4-one
C1C(=O)N(C(=S)S1)C2=CC=CC=C2
CAS Min % 97.0
CAS Max % 100.0
Melting Point 190°C to 194°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 209.29
Percent Purity 97+%
Physical Form Crystalline Powder
Chemical Name or Material N-Phenylrhodanine
Show More Show Less
compliance-icons
Product Identifier
  • N-Phenylrhodanine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'