missing translation for 'onlineSavingsMsg'
Learn More

N-Phenyliminodiacetic acid 97%, ACROS Organics™

Brand:  Acros Organics 391170250

Code : 29

Additional Details : CAS Number : 1137-73-1 Weight : 0.07500kg


Quantity 25g
Packaging Glass bottle

 View more versions of this product

Product Code. 10289623

  • £104.90 / 25g


Estimated Shipment
22-05-2018
Add to basket


Specifications

Chemical Identifiers

N-Phenyliminodiacetic acid
96.0
≥96.0%
C10H11NO4
209.2
N, N-Anilinediacetic acid
1137-73-1
100.0
97%
C6H5N(CH2COOH)2
MFCD00043570

Specifications

N-Phenyliminodiacetic acid
150° to 154°C
1137-73-1
100.0
Glass bottle
≥96.0%
C10H11NO4
209.2
N, N-Anilinediacetic acid
Grayish to beige
25g
96.0
Authentic
Powder
97%
C6H5N(CH2COOH)2
MFCD00043570
Videos
SDS