missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Phenylglycine ethyl ester, 99%, Thermo Scientific™
Click to view available options
Quantity:
5 g
50 g
Packaging:
Glass bottle
Specifications
CAS Min % | 98.5 |
CAS Max % | 100.0 |
Melting Point | 53.0°C to 56.0°C |
Color | White to Beige |
Infrared Spectrum | Authentic |
Assay Percent Range | 99% |
Packaging | Glass bottle |
Quantity | 5 g |
Merck Index | 15, 7403 |
Formula Weight | 179.22 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'