missing translation for 'onlineSavingsMsg'
Learn More

N-Phenylglycine ethyl ester, 99%, Thermo Scientific™

Catalog Number p-182729
Click to view available options
Quantity:
5 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
2216-92-4
C10H13NO2
179.22
MFCD00038315
MLSGRWDEDYJNER-UHFFFAOYSA-N
n-phenylglycine ethyl ester, ethyl phenylglycinate, ethyl 2-phenylamino acetate, ethyl n-phenylglycinate, glycine, n-phenyl-, ethyl ester, unii-7xl9t8bk0u, ethyl anilinoacetate, n-phenylglycineethylester, n-phenylglycine ethyl, 7xl9t8bk0u
75190
ethyl 2-anilinoacetate
CCOC(=O)CNC1=CC=CC=C1
This item is not returnable. View return policy
2216-92-4
C10H13NO2
179.22
MFCD00038315
MLSGRWDEDYJNER-UHFFFAOYSA-N
n-phenylglycine ethyl ester, ethyl phenylglycinate, ethyl 2-phenylamino acetate, ethyl n-phenylglycinate, glycine, n-phenyl-, ethyl ester, unii-7xl9t8bk0u, ethyl anilinoacetate, n-phenylglycineethylester, n-phenylglycine ethyl, 7xl9t8bk0u
75190
ethyl 2-anilinoacetate
CCOC(=O)CNC1=CC=CC=C1
CAS Min % 98.5
CAS Max % 100.0
Melting Point 53.0°C to 56.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 5 g
Merck Index 15, 7403
Formula Weight 179.22
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material N-Phenylglycine ethyl ester, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'