N-Phenylbenzohydroxamic acid, 98%, ACROS Organics™


Overview
Brand: Acros Organics

Manufacturer Part Number: 417371000

100GR N-Phenylbenzohydroxamic acid, 98%

UNSPSC: 12350000

Code: 29

Additional Details:
Additional Details: CAS Number: 304-88-1 Weight: 0.15000kg


Packaging Glass bottle
Quantity 100g

Product Code. 10669004

Quantity Price
1 £ 310.7 / 100g
EU Stock 1
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material N-Phenylbenzohydroxamic acid
CAS 304-88-1
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C13H11NO2
Linear Formula C6H5CON(OH)C6H5
Formula Weight 213.24
MDL Number MFCD00002111
Synonym N-Benzoyl-N-phenylhydroxylamine

Specification

Assay ≥97.5%
Physical Form Needle-Like Crystals or Crystalline Powder
Formula Weight 213.24
Color Light beige needle-like
Quantity 100g
CAS Min % 97.5
Linear Formula C6H5CON(OH)C6H5
CAS 304-88-1
Infrared Spectrum Authentic
Assay Percent Range 98%
MDL Number MFCD00002111
Packaging Glass bottle
Molecular Formula C13H11NO2
CAS Max % 100.0
Chemical Name or Material N-Phenylbenzohydroxamic acid
Synonym N-Benzoyl-N-phenylhydroxylamine
Melting Point 119° to 123°C
Documentation

Technical documents