missing translation for 'onlineSavingsMsg'
Learn More

N-Phenylbenzamidine, 97%, Thermo Scientific Chemicals

Catalog Number p-7021046
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
1527-91-9
C13H12N2
196.253
MFCD00019730
MPYOKHFSBKUKPQ-UHFFFAOYSA-N
n-phenylbenzamidine, n-phenylbenzenecarboximidamide, n'-phenylbenzamidine, n1-phenylbenzene-1-carboximidamide, n2-phenylbenzamidine, n-phenylbenzimidamide, acmc-20ak1f, maybridge1_004766, iminophenylmethyl phenylamine, n-phenyl-benzenecarboximidamide
200127
N'-phenylbenzenecarboximidamide
C1=CC=C(C=C1)C(=NC2=CC=CC=C2)N
This item is not returnable. View return policy
1527-91-9
C13H12N2
196.253
MFCD00019730
MPYOKHFSBKUKPQ-UHFFFAOYSA-N
n-phenylbenzamidine, n-phenylbenzenecarboximidamide, n'-phenylbenzamidine, n1-phenylbenzene-1-carboximidamide, n2-phenylbenzamidine, n-phenylbenzimidamide, acmc-20ak1f, maybridge1_004766, iminophenylmethyl phenylamine, n-phenyl-benzenecarboximidamide
200127
N'-phenylbenzenecarboximidamide
C1=CC=C(C=C1)C(=NC2=CC=CC=C2)N

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 117°C to 120°C
Quantity 5 g
Beilstein 608471
Sensitivity Hygroscopic
Formula Weight 196.25
Percent Purity 97%
Chemical Name or Material N-Phenylbenzamidine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'