missing translation for 'onlineSavingsMsg'
Learn More

N-Phenyl-p-phenylenediamine, 98%, Thermo Scientific Chemicals

Catalog Number 10696042
Click to view available options
Quantity:
100 g
250 g
1 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
101-54-2
C12H12N2
184.24
MFCD00007850
ATGUVEKSASEFFO-UHFFFAOYSA-N
4-aminodiphenylamine, n-phenyl-p-phenylenediamine, n1-phenylbenzene-1,4-diamine, p-anilinoaniline, azosalt r, p-semidine, p-aminodiphenylamine, luxan black r, semidine, n-phenyl-1,4-phenylenediamine
7564
CHEBI:59038
4-N-phenylbenzene-1,4-diamine
C1=CC=C(C=C1)NC2=CC=C(C=C2)N
This item is not returnable. View return policy
101-54-2
C12H12N2
184.24
MFCD00007850
ATGUVEKSASEFFO-UHFFFAOYSA-N
4-aminodiphenylamine, n-phenyl-p-phenylenediamine, n1-phenylbenzene-1,4-diamine, p-anilinoaniline, azosalt r, p-semidine, p-aminodiphenylamine, luxan black r, semidine, n-phenyl-1,4-phenylenediamine
7564
CHEBI:59038
4-N-phenylbenzene-1,4-diamine
C1=CC=C(C=C1)NC2=CC=C(C=C2)N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 69.0°C to 75.0°C
Boiling Point 354.0°C
Flash Point >250°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5NHC6H4NH2
Quantity 100 g
Beilstein 13, 76
Solubility Information Solubility in water: 0.6g/L (20°C). Other solubilities: soluble in alcohol and acetone
Formula Weight 184.24
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material N-Phenyl-p-phenylenediamine, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • N-Phenyl-p-phenylenediamine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'