missing translation for 'onlineSavingsMsg'
Learn More

N-Phenyl-p-phenylenediamine, 98%, ACROS Organics™

Brand:  Acros Organics 221000010

Code : 29

Additional Details : CAS Number : 101-54-2 Weight : 1.05000kg Transport : UN number : 3077 Chem class : 1 Pack group : III


Quantity 1kg
Packaging Glass bottle

 View more versions of this product

Product Code. 10490681

  • £140.60 / 1kg


Estimated Shipment
13-02-2018
Add to basket


Specifications

Chemical Identifiers

N-Phenyl-p-phenylenediamine
101-54-2
100.0
98%
C6H5NHC6H4NH2
MFCD00007850
98%
97.5
98%
C12H12N2
184.24
4-Aminodiphenylamine

Specifications

N-Phenyl-p-phenylenediamine
354.0°C
69.0° to 75.0°C
98%
Authentic
100.0
Liquid
98%
C6H5NHC6H4NH2
MFCD00007850
4-Aminodiphenylamine
>250°C
Clear
1kg
101-54-2
97.5
Glass bottle
98%
C12H12N2
184.24
13, 76
Solubility in water: 0.6g/L (20°C). Other solubilities: soluble in alcohol and acetone
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • N-Phenyl-p-phenylenediamine

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .

SDS