missing translation for 'onlineSavingsMsg'
Learn More

N-Phenyl-1-naphthylamine, 98%, Thermo Scientific Chemicals

Catalog Number 10635772
Click to view available options
Quantity:
25 g
250 g
Packaging:
Glass Bottle
90-30-2
C16H13N
219.29
MFCD00003878
XQVWYOYUZDUNRW-UHFFFAOYSA-N
n-phenyl-1-naphthylamine, 1-anilinonaphthalene, pana, antioxidant pan, n-1-naphthyl aniline, neozone a, 1-naphthalenamine, n-phenyl, phenylnaphthylamine, nonox a, aceto pan
7013
CHEBI:34876
N-phenylnaphthalen-1-amine
C1=CC=C(C=C1)NC2=CC=CC3=CC=CC=C32
90-30-2
C16H13N
219.29
MFCD00003878
XQVWYOYUZDUNRW-UHFFFAOYSA-N
n-phenyl-1-naphthylamine, 1-anilinonaphthalene, pana, antioxidant pan, n-1-naphthyl aniline, neozone a, 1-naphthalenamine, n-phenyl, phenylnaphthylamine, nonox a, aceto pan
7013
CHEBI:34876
N-phenylnaphthalen-1-amine
C1=CC=C(C=C1)NC2=CC=CC3=CC=CC=C32

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 58.0°C to 62.0°C
Color Brown to Pink
Boiling Point 226.0°C (15.0 mmHg)
Flash Point >200°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass Bottle
Linear Formula C10H7NHC6H5
Quantity 250 g
Beilstein 12, 1224
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol,ether and benzene
Formula Weight 219.29
Percent Purity 98%
Physical Form Crystals or Flakes
Chemical Name or Material N-Phenyl-1-naphthylamine, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • N-Phenyl-1-naphthylamine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'