missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ n-Pentylmagnesium bromide, 1M in THF

Grignard Reagent

Brand:  Alfa Aesar™ 041278.18

Code : NEW

Additional Details : CAS Number : 693-25-4 Weight : 0.05000kg Transport : UN number : 3399 Chem class : 1 Pack group : II

Quantity 50g

 View more versions of this product

Product Code. 11389657

  • £68.05 / 50g


Estimated Shipment date
from Supplier 15-02-2018
Add to basket

Specifications

Chemical Identifiers

n-Pentylmagnesium bromide
175.36
UN3399
693-25-4
MFCD00000045

Specifications

n-Pentylmagnesium bromide
0.98g/mL
Yellow to brown
693-25-4
175.36
UN3399
1M in THF
Air & Moisture Sensitive
50g
Liquid
MFCD00000045
Videos
SDS