missing translation for 'onlineSavingsMsg'
Learn More

n-Pentane, Spectrophotometric Grade, 99+% min., Thermo Scientific Chemicals

Catalog Number 11318387
Click to view available options
Quantity:
1 L
4 L
4 x 1 L
4 x 4 L
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
109-66-0
C5H12
72.15
MFCD00009498
OFBQJSOFQDEBGM-UHFFFAOYSA-N
n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish
8003
CHEBI:37830
pentane
CCCCC
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
109-66-0
C5H12
72.15
MFCD00009498
OFBQJSOFQDEBGM-UHFFFAOYSA-N
n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish
8003
CHEBI:37830
pentane
CCCCC

n-Pentane is used in spectrophotometry and environmental testing. It is used in consumer merchandise such as spot lifters/cleaners and foaming shave gels and in commercial products such as a blowing agent for expanded polystyrene (foam) insulation. It may also be used as a solvent (in pure form) in laboratory, chemical analysis, and other settings where a highly volatile non-polar solvent is needed.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point °C to 130°C
Color Colorless
Assay Percent Range >99%
Appearance Colorless liquid
Solubility Information Miscible with water.
Formula Weight 72.15
Grade Spectrophotometric
Density 1.626 g/cm3 at 20°C
Boiling Point 36°C
Quantity 4 x 4 L
Flash Point -49°C (-57°F)
Physical Form Liquid
Refractive Index 1.358
Vapor Pressure 554 hPa (416mm Hg) at 20°C
Chemical Name or Material n-Pentane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'