missing translation for 'onlineSavingsMsg'
Learn More

N-Pentane, Extra Pure, SLR, Fisher Chemical™

Catalog Number 10356000
Click to view available options
Quantity:
500 mL
2.5 L
5 L
25 L
Packaging:
Amber glass bottle
Metal can
Plastic Coated Glass bottle
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
109-66-0
C5H12
72.15
MFCD00009498
OFBQJSOFQDEBGM-UHFFFAOYSA-N
8003
CHEBI:37830
pentane
CCCCC
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
109-66-0
C5H12
72.15
MFCD00009498
OFBQJSOFQDEBGM-UHFFFAOYSA-N
8003
CHEBI:37830
pentane
CCCCC

N-Pentane, Extra Pure, SLR

TRUSTED_SUSTAINABILITY
Packaging Amber glass bottle
Quantity 500 mL
Formula Weight 72.15g/mol
Vapor Pressure 573mbar at 20°C
Viscosity 0.25 mPaS at 20°C
Vapor Density 2.5
Melting Point -130°C
Boiling Point 36°C
Physical Form Liquid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'