Learn More
N-Pentane, Extra Pure, SLR, Fisher Chemical™
Brand: Fisher Chemical P/1020/21
Code : 21
Additional Details : CAS Number : 109-66-0 Weight : 3.13000kg Transport : UN number : 1265 Chem class : 3 Pack group : II
Packaging | Metal can |
---|---|
Quantity | 5L |
Chemical Identifiers
109-66-0 | |
72.151 | |
OFBQJSOFQDEBGM-UHFFFAOYSA-N | |
CHEBI:37830 | |
CCCCC |
C5H12 | |
9498 | |
8003 | |
pentane |
Specifications
109-66-0 | |
9498 | |
CCCCC | |
72.151 | |
CHEBI:37830 | |
Liquid | |
573mbar at 20°C | |
2.5 | |
36°C | |
5L |
C5H12 | |
OFBQJSOFQDEBGM-UHFFFAOYSA-N | |
pentane | |
8003 | |
72.15g/mol | |
Metal can | |
0.25 mPaS at 20°C | |
-130°C | |
Undesignated |
Safety and Handling
- n-Pentane
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 2
- Aspiration toxicity Category 1
- Flammable liquid Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H304-May be fatal if swallowed and enters airways.
- H411-Toxic to aquatic life with long lasting effects.
- H336-May cause drowsiness or dizziness.
- H225-Highly flammable liquid and vapour.
- EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P331-Do NOT induce vomiting.