n-Pentane, for analysis, ACROS Organics™


Overview
Brand: GC Internal (Acros/Geel)

Manufacturer Part Number: 383900100

10LT n-Pentane, for analysis

UNSPSC: 12352100

Code: 50

Additional Details:
Additional Details: CAS Number: 109-66-0 Weight: 6.76000kg Transport: UN number : 1265 Chem class : 1 Pack group : II


Packaging Aluminium Can
Quantity 10L

Product Code. 10115653

Quantity Price
1 £ 176.1 / 10 L
Estimated Shipment
27-02-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material n-Pentane
CAS 109-66-0
Purity Grade For analysis
Assay ≥99.0%
Molecular Formula C5H12
Linear Formula CH3(CH2)3CH3
Formula Weight 72.15
MDL Number MFCD00009498

Specification

Assay ≥99.0%
Physical Form Liquid
Formula Weight 72.15
Quantity 10L
Color Clear
Merck Index 15, 7228
Packaging Aluminium Can
Silicon (Si) 0.02ppm max.
Sodium (Na) 1ppm max.
Trace Analysis 0.1ppm max.
Copper (Cu) 0.05ppm max.
Residue after Evaporation 0.002% max.
Linear Formula CH3(CH2)3CH3
Health Hazard 3 GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician. Do NOT induce vomiting. Avoid release to the environment. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Ground/Bond container and receiving equipment.
Magnesium (Mg) 0.1ppm max.
Purity Grade For analysis
CAS 109-66-0
Calcium (Ca) 0.5ppm max.
Acidity Alkalinity 0.0001 meq/g max.
Potassium (K) 0.2ppm max.
Total Sulfur 5ppm max.
MDL Number MFCD00009498
Iron (Fe) 0.2ppm max.
Total Phosphorus 0.5ppm max.
Water 0.01% max. (K.F.)
Health Hazard 2 GHS P Statement May be fatal if swallowed and enters airways. May cause drowsiness or dizziness. Toxic to aquatic life with long lasting effects. Highly flammable liquid and vapor. Repeated exposure may cause skin dryness or cracking.
Molecular Formula C5H12
Health Hazard 1 Danger
Density 0.6260g/mL
Chemical Name or Material n-Pentane
Lead (Pb) 0.05ppm max.
Documentation Safety and Handling

Danger

GHS P Statement May be fatal if swallowed and enters airways. May cause drowsiness or dizziness. Toxic to aquatic life with long lasting effects. Highly flammable liquid and vapor. Repeated exposure may cause skin dryness or cracking.

GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician. Do NOT induce vomiting. Avoid release to the environment. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Ground/Bond container and receiving equipment.