missing translation for 'onlineSavingsMsg'
Learn More

n-Octylamine, 99+%, Thermo Scientific Chemicals

Catalog Number p-4509851
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
111-86-4
IOQPZZOEVPZRBK-UHFFFAOYSA-N
octylamine, n-octylamine, 1-aminooctane, 1-octanamine, 1-octylamine, caprylamine, caprylylamine, armeen 8, n-octylamine, mono, armeen 8d
8143
CHEBI:7728
octan-1-amine
CCCCCCCCN
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
111-86-4
IOQPZZOEVPZRBK-UHFFFAOYSA-N
octylamine, n-octylamine, 1-aminooctane, 1-octanamine, 1-octylamine, caprylamine, caprylylamine, armeen 8, n-octylamine, mono, armeen 8d
8143
CHEBI:7728
octan-1-amine
CCCCCCCCN

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -1.0°C
Density 0.7820g/mL
Boiling Point 179.0°C
Flash Point 60°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Molecular Formula C8H19N
Linear Formula CH3(CH2)7NH2
Refractive Index 1.4280 to 1.4300
Quantity 5 g
Beilstein 04,196
Specific Gravity 0.782
Solubility Information Solubility in water: 0.2g/L water (25°C). Other solubilities: soluble in most common organic solvents
Molecular Weight (g/mol) 129.24
Viscosity 1.39 mPa.s (20°C)
Formula Weight 129.24
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material n-Octylamine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'