missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Octanoic Acid, Extra Pure, SLR, Fisher Chemical™
Click to view available options
Quantity:
500 mL
2.5 L
25 L
Packaging:
Amber glass bottle
Plastic Coated Glass bottle
Plastic container
Specifications
Melting Point | 16°C |
Color | Yellow |
Boiling Point | 237°C |
Packaging | Amber glass bottle |
Quantity | 500 mL |
Vapor Density | 5 |
Formula Weight | 144.21g/mol |
Physical Form | Liquid |
Safety and Handling
Product Identifier
- Octanoic acid
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 C
- H318-Causes serious eye damage.
- H412-Harmful to aquatic life with long lasting effects.
- H314-Causes severe skin burns and eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'