missing translation for 'onlineSavingsMsg'
Learn More
Learn More




N-Octane, Pure, SLR, Fisher Chemical™
Click to view available options
Quantity:
100mL
500mL
Specifications
Color | Colorless |
Melting Point | -57°C |
Boiling Point | 125°C |
Packaging | Amber glass bottle |
Quantity | 500mL |
Formula Weight | 114.23g/mol |
Physical Form | Liquid |
Vapor Density | 3.9 |
Safety and Handling




Product Identifier
- n-Octane
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Aspiration toxicity Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H304-May be fatal if swallowed and enters airways.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H315-Causes skin irritation.
- H336-May cause drowsiness or dizziness.
- H225-Highly flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P331-Do NOT induce vomiting.