missing translation for 'onlineSavingsMsg'
Learn More

n-Octane, 99+%, extra pure, Thermo Scientific Chemicals

Catalog Number p-3737102
Click to view available options
Quantity:
2 mL
2.5 L
25 L
Packaging:
Glass Bottle
Metal drum
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
111-65-9
C8H18
114.23
MFCD00009556
TVMXDCGIABBOFY-UHFFFAOYSA-N
n-octane, oktan, oktanen, ottani, oktanen dutch, heptane, methyl, oktan polish, ottani italian, octane, all isomers, unii-x1rv0b2fjv
356
CHEBI:17590
octane
CCCCCCCC
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
111-65-9
C8H18
114.23
MFCD00009556
TVMXDCGIABBOFY-UHFFFAOYSA-N
n-octane, oktan, oktanen, ottani, oktanen dutch, heptane, methyl, oktan polish, ottani italian, octane, all isomers, unii-x1rv0b2fjv
356
CHEBI:17590
octane
CCCCCCCC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 99.0
CAS Max % 100.0
Melting Point -57.0°C
Color Colorless
Density 0.7080g/mL
Boiling Point 125.0°C to 127.0°C
Flash Point 13°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass Bottle
Linear Formula CH3(CH2)6CH3
Refractive Index 1.3965 to 1.3985
Quantity 2 mL
Beilstein 01, 159
Residue after Evaporation 0.05% max.
Merck Index 15, 6838
Specific Gravity 0.708
Solubility Information Solubility in water: 0.0007g/L (20°C). Other solubilities: soluble in alcohol and acetone, freely miscible with ether, benzene, petroleum, ether and gasoline
Viscosity 0.55 mPa.s (20°C)
Formula Weight 114.23
Vapor Pressure 14mbar at 20°C
Percent Purity 99+%
Grade Extra Pure
Physical Form Liquid
Chemical Name or Material n-Octane
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • n-Octane
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Aspiration toxicity Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P273-Avoid release to the environment.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'