missing translation for 'onlineSavingsMsg'
Learn More

n-Octadecyl sulfate, sodium salt, 98%, Thermo Scientific Chemicals

Catalog Number p-3586463
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1120-04-3
C18H37NaO4S
372.55
MFCD00007469
NWZBFJYXRGSRGD-UHFFFAOYSA-M
sodium stearyl sulfate, octadecyl sulfate sodium salt, sodium octadecyl sulfate, octadecyl sodium sulfate, sulfuric acid, monooctadecyl ester, sodium salt, sodium octadecyl sulphate, sodium monostearyl sulfate, unii-t4eww0y4ej, sodium n-octadecyl sulfate, sodium monooctadecyl sulfate
23673604
sodium;octadecyl sulfate
CCCCCCCCCCCCCCCCCCOS(=O)(=O)[O-].[Na+]
This item is not returnable. View return policy
1120-04-3
C18H37NaO4S
372.55
MFCD00007469
NWZBFJYXRGSRGD-UHFFFAOYSA-M
sodium stearyl sulfate, octadecyl sulfate sodium salt, sodium octadecyl sulfate, octadecyl sodium sulfate, sulfuric acid, monooctadecyl ester, sodium salt, sodium octadecyl sulphate, sodium monostearyl sulfate, unii-t4eww0y4ej, sodium n-octadecyl sulfate, sodium monooctadecyl sulfate
23673604
sodium;octadecyl sulfate
CCCCCCCCCCCCCCCCCCOS(=O)(=O)[O-].[Na+]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 211.0°C to 213.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Ex Sulfated Ash)
Packaging Glass bottle
Linear Formula CH3(CH2)17OSO3Na
Quantity 1 g
Solubility Information Solubility in water: soluble.
Formula Weight 372.55
Percent Purity 98%
Physical Form Powder
Chemical Name or Material n-Octadecyl sulfate, sodium salt
Show More Show Less
compliance-icons
Product Identifier
  • n-Octadecyl sulfate, sodium salt
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'