missing translation for 'onlineSavingsMsg'
Learn More

n-Octadecane, tech., 90%, Thermo Scientific Chemicals

Catalog Number p-7059247
Click to view available options
Quantity:
1 kg
5 kg
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
593-45-3
C18H38
254.502
MFCD00009007
RZJRJXONCZWCBN-UHFFFAOYSA-N
n-octadecane, octadecan, unii-n102p6haiu, n102p6haiu, octadecane, octadecane, analytical standard, octadecane, n, oktadekan, ccris 681, stearyl group
11635
CHEBI:32926
octadecane
CCCCCCCCCCCCCCCCCC
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
593-45-3
C18H38
254.502
MFCD00009007
RZJRJXONCZWCBN-UHFFFAOYSA-N
n-octadecane, octadecan, unii-n102p6haiu, n102p6haiu, octadecane, octadecane, analytical standard, octadecane, n, oktadekan, ccris 681, stearyl group
11635
CHEBI:32926
octadecane
CCCCCCCCCCCCCCCCCC

n-octadecane is used as a solvent, a lubricant, transformer oil and an anti-corrosion agent. It is also used in paraffin and as a chemical intermediate in organic synthesis. It plays an important role in pheromones as a chemical messenger. N-Octadecane/ molecular sieve 5A composites find application as thermal storage material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 28°C
Color White
Density 0.782 g/mL
Boiling Point 317°C
Flash Point 166°C (330°F)
Odor Odorless
Refractive Index 1.439
Quantity 1 kg
Solubility Information Miscible with alcohol,acetone,ether and petroleum ether. Immiscible with water.
Formula Weight 254.5
Percent Purity 90%
Grade Technical
Physical Form Crystalline Mass/Melt
Chemical Name or Material n-Octadecane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'