missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ n-Octadecane, 99.5+%

Brand:  Alfa Aesar™ 044512.14

Code : NEW

Additional Details : CAS Number : 593-45-3 Weight : 0.02500kg

Quantity 25g

 View more versions of this product

Product Code. 11485240

  • £112.78 / 25g


Estimated Shipment date
from Supplier 06-06-2018
Add to basket

Specifications

Chemical Identifiers

n-Octadecane
CH3(CH2)16CH3
MFCD00009007
593-45-3
254.5

Specifications

n-Octadecane
1.4390
28°C
White
≥99.5%
166°C (330°F)
Odorless
MFCD00009007
593-45-3
0.782g/mL
317°C
Crystalline Mass/Melt
25g
CH3(CH2)16CH3
254.5
Videos
SDS