missing translation for 'onlineSavingsMsg'
Learn More

N,O-Dimethylhydroxylamine hydrochloride, 98%, ACROS Organics™

£23.21 - £193.00

Chemical Identifiers

CAS 6638-79-5
Molecular Formula C2H7NO·HCl
Molecular Weight (g/mol) 97.55
MDL Number MFCD00012485
InChI Key USZLCYNVCCDPLQ-UHFFFAOYSA-N
Synonym n,o-dimethylhydroxylamine hydrochloride, n,o-dimethylhydroxylamine hcl, n-methoxymethylamine hydrochloride, methanamine, n-methoxy-, hydrochloride, o,n-dimethylhydroxylamine hydrochloride, n-methoxymethanamine hydrochloride, o,n-dimethyl-hydroxylamine hydrochloride, methoxy methyl amine hydrochloride, n,o-dimethylhydroxyamine hydrochloride, n,o-dimethylhydroxylaminehydrochloride
PubChem CID 81138
IUPAC Name N-methoxymethanamine;hydrochloride
SMILES CNOC.Cl
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10570741
SDS View Documents
Acros Organics
116340050
5g Glass bottle
£23.21
5g
View alternative products
Save Up to 
»
In stock, ready to ship: 5 Please to purchase this item. Need a web account? Register with us today!
10030790
SDS View Documents
Acros Organics
116340250
25g Glass bottle
£71.73
25g
View alternative products
Save Up to 
»
In stock, ready to ship: 3 Please to purchase this item. Need a web account? Register with us today!
10316420
SDS View Documents
Acros Organics
116341000
100g Glass bottle
£193.00
100g
View alternative products
Save Up to 
»
In stock, ready to ship: 2 Please to purchase this item. Need a web account? Register with us today!
Description

Description

Coupling reagent
Specifications

Chemical Identifiers

6638-79-5
97.55
USZLCYNVCCDPLQ-UHFFFAOYSA-N
81138
CNOC.Cl
C2H7NO·HCl
MFCD00012485
n,o-dimethylhydroxylamine hydrochloride, n,o-dimethylhydroxylamine hcl, n-methoxymethylamine hydrochloride, methanamine, n-methoxy-, hydrochloride, o,n-dimethylhydroxylamine hydrochloride, n-methoxymethanamine hydrochloride, o,n-dimethyl-hydroxylamine hydrochloride, methoxy methyl amine hydrochloride, n,o-dimethylhydroxyamine hydrochloride, n,o-dimethylhydroxylaminehydrochloride
N-methoxymethanamine;hydrochloride

Specifications

(10% aq. soln.) clear
Authentic
0.5% max.
112.0°C to 116.0°C
98%
97.5
98%
CH3ONHCH3·HCl
04, 534
n,o-dimethylhydroxylamine hydrochloride, n,o-dimethylhydroxylamine hcl, n-methoxymethylamine hydrochloride, methanamine, n-methoxy-, hydrochloride, o,n-dimethylhydroxylamine hydrochloride, n-methoxymethanamine hydrochloride, o,n-dimethyl-hydroxylamine hydrochloride, methoxy methyl amine hydrochloride, n,o-dimethylhydroxyamine hydrochloride, n,o-dimethylhydroxylaminehydrochloride
USZLCYNVCCDPLQ-UHFFFAOYSA-N
N-methoxymethanamine;hydrochloride
81138
Crystalline Powder or Crystals
N, O-Dimethylhydroxylamine hydrochloride
Glass bottle
White
5g
6638-79-5
100.0
C2H7NO·HCl
MFCD00012485
11,201; 12,205; 15,144
(10% in water) Clear
CNOC.Cl
97.55
97.55
98%
Safety and Handling

Safety and Handling

Product Identifier
  • N,O-Dimethylhydroxylamine hydrochloride

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS