missing translation for 'onlineSavingsMsg'
Learn More

N,O-Di-BOC-hydroxylamine, 97%, Thermo Scientific Chemicals

Catalog Number p-4090066
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
85006-25-3
C10H19NO5
233.26
MFCD00034797
AGOSGCWATIJZHQ-UHFFFAOYSA-N
tert-butyl tert-butoxycarbonyl oxycarbamate, n,o-di-boc-hydroxylamine, tert-butyl n-tert-butoxycarbonyloxy carbamate, n,o-bis tert-butoxycarbonyl hydroxylamine, tert-butyl tert-butoxycarbonyl oxy carbamate, tert-butyl 1,1-dimethylethoxy carbonyl oxy carbamate, tert-butyl tert-butoxycarbonyloxy carbamate, tert-butoxycarbonylamino tert-butyl carbonate, tert-butyl 2-methylpropan-2-yl oxycarbonylamino carbonate, 2-tert-butoxy carbonyl amino oxy carbonyl oxy-2-methylpropane
2728863
tert-butyl [(2-methylpropan-2-yl)oxycarbonylamino] carbonate
CC(C)(C)OC(=O)NOC(=O)OC(C)(C)C
This item is not returnable. View return policy
85006-25-3
C10H19NO5
233.26
MFCD00034797
AGOSGCWATIJZHQ-UHFFFAOYSA-N
tert-butyl tert-butoxycarbonyl oxycarbamate, n,o-di-boc-hydroxylamine, tert-butyl n-tert-butoxycarbonyloxy carbamate, n,o-bis tert-butoxycarbonyl hydroxylamine, tert-butyl tert-butoxycarbonyl oxy carbamate, tert-butyl 1,1-dimethylethoxy carbonyl oxy carbamate, tert-butyl tert-butoxycarbonyloxy carbamate, tert-butoxycarbonylamino tert-butyl carbonate, tert-butyl 2-methylpropan-2-yl oxycarbonylamino carbonate, 2-tert-butoxy carbonyl amino oxy carbonyl oxy-2-methylpropane
2728863
tert-butyl [(2-methylpropan-2-yl)oxycarbonylamino] carbonate
CC(C)(C)OC(=O)NOC(=O)OC(C)(C)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 67°C to 70°C
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 233.26
Percent Purity 97%
Chemical Name or Material N, O-Di-BOC-hydroxylamine
compliance-icons
Product Identifier
  • N,O-Di-BOC-hydroxylamine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'