missing translation for 'onlineSavingsMsg'
Learn More

N,O-Bis(trimethylsilyl)trifluoroacetamide, with 1% TMCS, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals

Catalog Number p-7059076
Click to view available options
Quantity:
5 g
25 g
25561-30-2
C8H18F3NOSi2
257.403
MFCD00008269
XCOBLONWWXQEBS-GHXNOFRVSA-N
bstfa, n,o-bis trimethylsilyl trifluoroacetamide, acetamide, 2,2,2-trifluoro-o,n-bis trimethylsilyl, trimethylsilyl 2,2,2-trifluoro-n-trimethylsilyl acetimidate, trimethylsilyl 2,2,2-trifluoro-n-trimethylsilyl ethanimidate, trimethylsilyl 1z-2,2,2-trifluoro-n-z-trimethylsilyl ethanimidoate #, n,o-bis trimethylsilyl trifluoroacetamide bstfa, trimethylsilyl 2,2,2-trifluoro-n-trimethylsilyl ethanecarboximidate
9601896
trimethylsilyl (1Z)-2,2,2-trifluoro-N-trimethylsilylethanimidate
C[Si](C)(C)N=C(C(F)(F)F)O[Si](C)(C)C
25561-30-2
C8H18F3NOSi2
257.403
MFCD00008269
XCOBLONWWXQEBS-GHXNOFRVSA-N
bstfa, n,o-bis trimethylsilyl trifluoroacetamide, acetamide, 2,2,2-trifluoro-o,n-bis trimethylsilyl, trimethylsilyl 2,2,2-trifluoro-n-trimethylsilyl acetimidate, trimethylsilyl 2,2,2-trifluoro-n-trimethylsilyl ethanimidate, trimethylsilyl 1z-2,2,2-trifluoro-n-z-trimethylsilyl ethanimidoate #, n,o-bis trimethylsilyl trifluoroacetamide bstfa, trimethylsilyl 2,2,2-trifluoro-n-trimethylsilyl ethanecarboximidate
9601896
trimethylsilyl (1Z)-2,2,2-trifluoro-N-trimethylsilylethanimidate
C[Si](C)(C)N=C(C(F)(F)F)O[Si](C)(C)C

N,O-Bis(trimethylsilyl)trifluoroacetamide is used for analytical purposes or as a chemical reagent for synthesis of more complex molecules. It is silylation reagent useful in GC and GC-MS for the derivatization of a wide range of functional groups under mild conditions. It is also used as DAT inhibitor and a SLC6A2 inhibitor. It is routinely used to derivatize carboxylic acids, phenols, steroids, amines, alcohols, and alkaloids and amides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
TRUSTED_SUSTAINABILITY
Melting Point -10°C
Color Colorless to Yellow
Density 0.97 g/mL
Boiling Point 145°C to 147°C
Flash Point 23°C (73°F)
Packaging ChemSeal™ bottles
Refractive Index 1.384
Quantity 5 g
UN Number UN2924
Beilstein 3607620
Sensitivity Moisture sensitive
Solubility Information Soluble in water (hydrolyses),chloroform and hexane.
Formula Weight 257.4
Physical Form Liquid
Chemical Name or Material N,O-Bis (trimethylsilyl)trifluoroacetamide, With 1% TMCS
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'