missing translation for 'onlineSavingsMsg'
Learn More

N,O-Bis(trimethylsilyl)hydroxylamine, 97%, ACROS Organics™

Brand:  Acros Organics 215980050

Code : 29

Additional Details : CAS Number : 22737-37-7 Weight : 0.05500kg Transport : UN number : 2920 Chem class : 1a Pack group : II


Quantity 5g
Packaging Glass bottle

 View more versions of this product

Product Code. 10451831

  • £60.30 / 5g


EU Stock 2
Add to basket


Specifications

Chemical Identifiers

N, O-Bis(trimethylsilyl)hydroxylamine
22737-37-7
100.0
97%
(CH3)3SiONHSi(CH3)3
MFCD00008266
97%
96.0
97%
C6H19NOSi2
177.4

Specifications

N, O-Bis(trimethylsilyl)hydroxylamine
22737-37-7
100.0
Authentic
Liquid
97%
C6H19NOSi2
177.4
06,58
78.0° to 80.0°C (100.0 mmHg)
28°C
97%
96.0
0.8300g/mL
Glass bottle
97%
1.4112
(CH3)3SiONHSi(CH3)3
MFCD00008266
Clear colorless
5g
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • N,O-Bis(trimethylsilyl)hydroxylamine

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS