missing translation for 'onlineSavingsMsg'
Learn More

N,O-Bis(trimethylsilyl)acetamide, 95%, Thermo Scientific Chemicals

Catalog Number 10564454
Click to view available options
Quantity:
25 mL
100 mL
1 L
Packaging:
Glass bottle
10416-59-8
C8H21NOSi2
203.43
MFCD00008270
SIOVKLKJSOKLIF-UHFFFAOYSA-N
n,o-bis trimethylsilyl acetamide, trimethylsilyl 1z-n-trimethylsilylethanimidate
6913588
CC(O[Si](C)(C)C)=N[Si](C)(C)C
10416-59-8
C8H21NOSi2
203.43
MFCD00008270
SIOVKLKJSOKLIF-UHFFFAOYSA-N
n,o-bis trimethylsilyl acetamide, trimethylsilyl 1z-n-trimethylsilylethanimidate
6913588
CC(O[Si](C)(C)C)=N[Si](C)(C)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Silylation reagent
TRUSTED_SUSTAINABILITY
CAS Min % 94.0
CAS Max % 100.0
Melting Point -8°C
Color Beige to Brown or Yellow
Density 0.8200g/mL
Boiling Point 71°C to 73°C (35.0 mmHg)
Flash Point 42°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula CH3C[=NSi(CH3)3]OSi(CH3)3
Refractive Index 1.4165 to 1.4185
Quantity 1 L
Fieser 01,61; 02,30; 03,23; 13,34
Specific Gravity 0.82
Solubility Information Solubility in water: decomposes
IUPAC Name trimethylsilyl (1E)-N-trimethylsilylethanimidate
Formula Weight 203.43
Percent Purity 95%
Physical Form Crystalline Powder, Crystals or Chunks
Chemical Name or Material N, O-Bis(trimethylsilyl)acetamide
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • N,O-Bis(trimethylsilyl)acetamide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • H226-Flammable liquid and vapour.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'