missing translation for 'onlineSavingsMsg'
Learn More

n-Nonanoic acid, 97%, Thermo Scientific Chemicals

Catalog Number 10762495
Click to view available options
Quantity:
100 g
500 g
1 kg
2.5 kg
Packaging:
Glass bottle
Plastic Drum
This item is not returnable. View return policy
112-05-0
C9H18O2
158.24
MFCD00004433
FBUKVWPVBMHYJY-UHFFFAOYSA-N
pelargonic acid, n-nonanoic acid, nonylic acid, pelargic acid, nonoic acid, n-nonylic acid, n-nonoic acid, 1-octanecarboxylic acid, pelargon, cirrasol 185a
8158
CHEBI:29019
nonanoic acid
CCCCCCCCC(=O)O
This item is not returnable. View return policy
112-05-0
C9H18O2
158.24
MFCD00004433
FBUKVWPVBMHYJY-UHFFFAOYSA-N
pelargonic acid, n-nonanoic acid, nonylic acid, pelargic acid, nonoic acid, n-nonylic acid, n-nonoic acid, 1-octanecarboxylic acid, pelargon, cirrasol 185a
8158
CHEBI:29019
nonanoic acid
CCCCCCCCC(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 12.5°C
Color Colorless
Density 0.9000g/mL
Boiling Point 254.0°C
Flash Point 114°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)7CO2H
Refractive Index 1.4299 to 1.4339
Quantity 500 g
Beilstein 02, 352
Merck Index 15, 7179
Specific Gravity 0.9
Solubility Information Solubility in water: negligible. Other solubilities: soluble in alcohol, chloroform and ether, soluble in most other commmon organic solvents
Formula Weight 158.24
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material n-Nonanoic acid
Show More Show Less
compliance-icons
Product Identifier
  • n-Nonanoic acid
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H412-Harmful to aquatic life with long lasting effects.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'