missing translation for 'onlineSavingsMsg'
Learn More

n-Nitrosomorpholine, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Brand:  SPEX Certiprep S-2749

Additional Details : Weight : 0.01000kg

Packaging 2mL amber prescored glass ampule
Quantity 1mL

Product Code. 16319377

  • £29.09 / 1mL

Estimated Shipment
22-07-2020

Please to purchase this item. Need a web account? Register with us today!


Includes: Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Description

Description

AnalyteCAS No.Concentration
N-Nitrosomorpholine59-89-21000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Specifications

n-Nitrosomorpholine
99.9
C4H8N2O2
C1COCCN1N=O
116.12
CHEBI:76326
Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
1mL
67-56-1
99.9
ZKXDGKXYMTYWTB-UHFFFAOYSA-N
4-nitrosomorpholine
6046
2mL amber prescored glass ampule
Methanol-P&T
GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
Videos
Documents

Product intended for laboratory use only