missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-n-Propyl-5,6,7,8-tetrahydronaphthalene-1-carboxamide, 95%

Brand:  Alfa Aesar™ H65133.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15469396

  • £74.73 / 1g


Estimated Shipment date
from Supplier 08-05-2018
Add to basket

Specifications

Chemical Identifiers

N-n-Propyl-5,6,7,8-tetrahydronaphthalene-1-carboxamide
217.31
MFCD27976897

Specifications

N-n-Propyl-5,6,7,8-tetrahydronaphthalene-1-carboxamide
Solid
217.31
1g
95%
MFCD27976897
Videos
SDS