missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-n-Pentyl-5,6,7,8-tetrahydronaphthalene-1-carboxamide, 95%

Brand:  Alfa Aesar™ H65911.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15469996

  • £59.78 / 1g


Estimated Shipment date
from Supplier 06-06-2018
Add to basket

Specifications

Chemical Identifiers

N-n-Pentyl-5,6,7,8-tetrahydronaphthalene-1-carboxamide
245.37
MFCD27976900

Specifications

N-n-Pentyl-5,6,7,8-tetrahydronaphthalene-1-carboxamide
Solid
245.37
1g
95%
MFCD27976900
Videos
SDS