missing translation for 'onlineSavingsMsg'
Learn More

N,N,N',N'-Tetraphenylbenzidine, 97%, Thermo Scientific Chemicals

Catalog Number 15426038
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
15546-43-7
C36H28N2
488.63
MFCD00228123
DCZNSJVFOQPSRV-UHFFFAOYSA-N
n,n,n',n'-tetraphenylbenzidine, tetraphenylbenzidine, n4,n4,n4',n4'-tetraphenyl-1,1'-biphenyl-4,4'-diamine, n,n,n,n-tetraphenylbenzidine, 1,1'-biphenyl-4,4'-diamine, n,n,n',n'-tetraphenyl, n,n'-diphenyl-n,n',di-m-tolyl-benzidine, n,n,n',n'-tetraphenyl-1,1'-biphenyl-4,4'-diamine, 4,4'-bis diphenylamino biphenyl, 1,1'-biphenyl-4,4'-diamine, n4,n4,n4',n4'-tetraphenyl
84981
N,N-diphenyl-4-[4-(N-phenylanilino)phenyl]aniline
C1=CC=C(C=C1)N(C1=CC=CC=C1)C1=CC=C(C=C1)C1=CC=C(C=C1)N(C1=CC=CC=C1)C1=CC=CC=C1
This item is not returnable. View return policy
15546-43-7
C36H28N2
488.63
MFCD00228123
DCZNSJVFOQPSRV-UHFFFAOYSA-N
n,n,n',n'-tetraphenylbenzidine, tetraphenylbenzidine, n4,n4,n4',n4'-tetraphenyl-1,1'-biphenyl-4,4'-diamine, n,n,n,n-tetraphenylbenzidine, 1,1'-biphenyl-4,4'-diamine, n,n,n',n'-tetraphenyl, n,n'-diphenyl-n,n',di-m-tolyl-benzidine, n,n,n',n'-tetraphenyl-1,1'-biphenyl-4,4'-diamine, 4,4'-bis diphenylamino biphenyl, 1,1'-biphenyl-4,4'-diamine, n4,n4,n4',n4'-tetraphenyl
84981
N,N-diphenyl-4-[4-(N-phenylanilino)phenyl]aniline
C1=CC=C(C=C1)N(C1=CC=CC=C1)C1=CC=C(C=C1)C1=CC=C(C=C1)N(C1=CC=CC=C1)C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 230°C to 234°C
Quantity 1 g
Formula Weight 488.62
Percent Purity 97%
Physical Form Solid
Chemical Name or Material N,N,N',N'-Tetraphenylbenzidine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'