missing translation for 'onlineSavingsMsg'
Learn More

N,N,N',N'-Tetramethyl-S-(1-oxido-2-pyridyl)thiuronium hexafluorophosphate, 97%, Thermo Scientific™

Catalog Number 11936401
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
212333-72-7
C10H16N3OS·F6P
371.29
UCOGEMMJHLHOAW-UHFFFAOYSA-N
hott, n,n,n',n'-tetramethyl-s-1-oxido-2-pyridyl thiuronium hexafluorophosphate, n,n,n',n'-tetramethyl-s-1-oxido-2-pyridyl thiouronium hexafluorophosphate, hott n,n,n',n'-tetramethyl-s-1-oxido-2-pyridyl thiuronium hexafluorophosphate, ambotzrl-1156, acmc-209fij, ksc911c8f, n,n,n',n'-tetramethyl-s-1-oxido-2-pyridyl thiuron, n,n,n',n'-tetramethyl-s-1-oxido-2-pyridyl thiuronhexafluorophosphate, n,n,n,n-tetramethyl-s-1-oxido-2-pyridyl thiouronium hexafluorophosphate
10571248
[dimethylamino-(1-oxidopyridin-1-ium-2-yl)sulfanylmethylidene]-dimethylazanium;hexafluorophosphate
CN(C)C(=[N+](C)C)SC1=CC=CC=[N+]1[O-].F[P-](F)(F)(F)(F)F
This item is not returnable. View return policy
212333-72-7
C10H16N3OS·F6P
371.29
UCOGEMMJHLHOAW-UHFFFAOYSA-N
hott, n,n,n',n'-tetramethyl-s-1-oxido-2-pyridyl thiuronium hexafluorophosphate, n,n,n',n'-tetramethyl-s-1-oxido-2-pyridyl thiouronium hexafluorophosphate, hott n,n,n',n'-tetramethyl-s-1-oxido-2-pyridyl thiuronium hexafluorophosphate, ambotzrl-1156, acmc-209fij, ksc911c8f, n,n,n',n'-tetramethyl-s-1-oxido-2-pyridyl thiuron, n,n,n',n'-tetramethyl-s-1-oxido-2-pyridyl thiuronhexafluorophosphate, n,n,n,n-tetramethyl-s-1-oxido-2-pyridyl thiouronium hexafluorophosphate
10571248
[dimethylamino-(1-oxidopyridin-1-ium-2-yl)sulfanylmethylidene]-dimethylazanium;hexafluorophosphate
CN(C)C(=[N+](C)C)SC1=CC=CC=[N+]1[O-].F[P-](F)(F)(F)(F)F
Coupling reagent
TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 111.7°C to 113.6°C
Color White to Yellow
Elemental Analysis 30.41 to 34.29% C, 10.64 to 12.00% N
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass Bottle
Quantity 5 g
Formula Weight 371.29
Percent Purity 97%
Physical Form Powder
Chemical Name or Material N,N,N',N'-Tetramethyl-S-(1-oxido-2-pyridyl)thiuronium hexafluorophosphate
Show More Show Less
compliance-icons
Product Identifier
  • N,N,N',N'-Tetramethyl-S-(1-oxido-2-pyridyl)thiuronium hexafluorophosphate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'